Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals TRIS-buffered saline (TBS, 10X), with 0.5% Tween 20

Catalog No. AAJ60448K2
Change view
Click to view available options
Quantity:
500 mL
1 L
2 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60448K2 1 L
AAJ60448AP 500 mL
AAJ60448K3 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ60448K2 Supplier Thermo Scientific Chemicals Supplier No. J60448K2
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol

TRIS-buffered saline (TBS, 10X), with 0.5% Tween 20 is used as a wash buffer and for sample preparation in general immunoassay applications, Tween 20 reduces nonspecific binding and protein:protein interactions.TBS is often used for diluting secondary antibodies or streptavidin-AP conjugate. TBS-Tween 20 is often used for washing steps.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TRIS-buffered saline (TBS, 10X), with 0.5% Tween 20 is used as a wash buffer and for sample preparation in general immunoassay applications, Tween 20 reduces nonspecific binding and protein:protein interactions.TBS is often used for diluting secondary antibodies or streptavidin-AP conjugate. TBS-Tween 20 is often used for washing steps.

Solubility
It is soluble in water.

Notes
250mM Tris, 27mM KCl, 1.37M NaCl, pH 7.4 with 0.5% Tween 20
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00678198
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Color Colorless
Concentration 10X
Physical Form Viscous Liquid
Quantity 1 L
Solubility Information It is soluble in water.
Chemical Name or Material TRIS-buffered saline, with 0.5% Tween 20
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.