Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals TRIS-buffered saline (TBS, 20X) pH 7.4 PROMO

Catalog No. J60877K3F25
Change view
Click to view available options
2 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
J60877K3F25 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. J60877K3F25 Supplier Thermo Scientific Chemicals Supplier No. J60877K3PR/DEL

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

View Alternate Product

Tris Buffered Saline (TBS) is a pH stabilizing solution used for western blot and ELISA procedures.

  • Applications: Used as a convenient prepared buffer for use in various applications.
  • Solubility: It is soluble in water.
  • Contains: 0.5 M Tris, 54 mM KCl, and 2.74 M NaCl, pH 7.4
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11 NO3
Molecular Weight (g/mol) 121.136 g/mol
MDL Number MFCD00132476
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Concentration 20X
pH 7.4
Physical Form Liquid
Quantity 2 L
Solubility Information Soluble in Water
Chemical Name or Material TRIS-buffered Saline
TSCA Yes
Recommended Storage Ambient Temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.