Learn More
Polysciences, Inc. Tris Buffered Saline; Composition: 1M Tris-HCl, 3M Sodium chloride; 8.0+/-0.2 pH; Storage: 25 deg.C
SDP
Catalog No. NC9948272
Encompass
Tris Buffered Saline; Composition: 1M Tris-HCl, 3M Sodium chloride; 8.0+/-0.2 pH; Storage: 25 deg.C
Catalog No. NC9948272 Supplier Polysciences, Inc. Supplier No. 240821
Tris Buffered Saline; Composition: 1M Tris-HCl, 3M Sodium chloride; 8.0+/-0.2 pH; Storage: 25 deg.C