Learn More
Santa Cruz Biotechnology Tris Buffered Saline with Tween 20: 5 L of 20X, 5 L
Catalog No. NC1955662
Encompass
Tris Buffered Saline with Tween 20: 5 L of 20X is a high-quality 20X concentrate for dilution to a 1X working solution commonly used as a wash solution for western blot membranes and microtiter plate wells in ELISA assays. Tris Buffered Saline with Tween 20 (TBST) is an optimal formulation of pH stabilizers, salts and detergents designed to effectively remove excess material from the microtiter plate wells without disrupting the ELISA binding reaction. By maintaining the proper buffering environment, unbound components can be washed away without suppressing antigen-antibody binding interactions, thereby reducing nonspecific background and increasing the specific signal.
Catalog No. NC1955662 Supplier Santa Cruz Biotechnology Supplier No. SC362311
Tris Buffered Saline with Tween 20: 5 L of 20X is a high-quality 20X concentrate for dilution to a 1X working solution commonly used as a wash solution for western blot membranes and microtiter plate wells in ELISA assays. Tris Buffered Saline with Tween 20 (TBST) is an optimal formulation of pH stabilizers, salts and detergents designed to effectively remove excess material from the microtiter plate wells without disrupting the ELISA binding reaction. By maintaining the proper buffering environment, unbound components can be washed away without suppressing antigen-antibody binding interactions, thereby reducing nonspecific background and increasing the specific signal.