Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp RPI TRIS-CAPS Buffer 10X Solution, 2 x 500 Milliliter Bottles
SDP

Catalog No. 501978317
Encompass_Preferred

This product is shipped with one 500 ml Anode bottle and one 500 ml Cathode bottle

10X Anode Formulation:

pH: 9.6
TRIS: 0.6 M
CAPS: 0.4M

10X Cathode Formulation:
pH: 9.5
TRIS: 0.6 M
CAPS: 0.4 M
SDS: 1%

Catalog No. 50-197-8317 Supplier Research Products International Corp Supplier No. T275001000.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is shipped with one 500 ml Anode bottle and one 500 ml Cathode bottle

10X Anode Formulation:

pH: 9.6
TRIS: 0.6 M
CAPS: 0.4M

10X Cathode Formulation:
pH: 9.5
TRIS: 0.6 M
CAPS: 0.4 M
SDS: 1%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.