Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris, Cell Culture Reagent, ≥99.95%, MP Biomedicals™

Catalog No. 0219455794 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-194-55794 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-194-55794 Supplier MP Biomedicals, Inc Supplier No. 0219455794
Add to Cart
Add to Cart

Tris, Cell Culture Reagent, ≥99.95% is a biological buffer for various applications.

  • Tris is an excellent biochemical and biological buffer for all applications where ultra high purity is required.
  • The pH values of all buffers are temperature and concentration dependent.
  • For Tris buffers, pH increases about 0.03 unit per °C decrease in temperature, and decreases 0.03 to 0.05 unit per ten-fold dilution.
  • For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.
TRUSTED_SUSTAINABILITY

Specifications

Absorbance OD260 nm (5% aq. soln.) ≤0.1
CAS 77-86-1
Melting Point 171 to 172°C
Boiling Point 219 to 220°C at 10 mm Hg
pH 10.4 (0.1 molar aq. soln.)
Physical Form Crystalline Powder
Quantity 5 kg
Vapor Pressure 2.2X10-5 mm Hg at 25°C
Synonym Trometamol, Tromethamine, Tris-[hydroxymethyl]aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base
Solubility Information 79.1 mg/mL in ethylene glycol, 26 mg/mL in methanol, 14.6 mg/mL in anhyd ethanol, 22 mg/mL in 95% ethanol, 14 mg/mL in dimethyl formamide, 20 mg/mL in acetone, 0.5 mg/mL in ethyl acetate, 0.4 mg/mL in olive oil, 0.1 mg/mL in cyclohexane, 0.05 mg/mL in chloroform, less than 0.05 mg/mL in carbon tetrachloride
Molecular Weight (g/mol) 121.136 g/mol
Percent Purity ≥95%
Grade Cell Culture Grade
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.