Learn More
Strem, An Ascensus Company CAS# 52522-40-4. Tris(dibenzylideneacetone)dipalladium(0) chloroform adduct. MFCD00075479. 2G
SDP
Catalog No. 5090112483
Encompass
CAS# 52522-40-4. 2G. Tris(dibenzylideneacetone)dipalladium(0) chloroform adduct. MFCD00075479. Molecular Formula: C52H43Cl3O3Pd2. Molecular Weight: 1035.08. Color/Form: red to brown solid. Stability: air sensitive, moisture sensitive. Strem#: 46-3010 www.strem.com/catalog/v3/46-3010/
Catalog No. 50-901-12483 Supplier Strem, An Ascensus Company Supplier No. 4630102G
CAS# 52522-40-4. 2G. Tris(dibenzylideneacetone)dipalladium(0) chloroform adduct. MFCD00075479. Molecular Formula: C52H43Cl3O3Pd2. Molecular Weight: 1035.08. Color/Form: red to brown solid. Stability: air sensitive, moisture sensitive. Strem#: 46-3010 www.strem.com/catalog/v3/46-3010/