Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tris(Dibenzylideneacetone)Dipalladium(0)-Chloroform Adduct

Catalog No. 366315250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-631-5250MG 250 mg
36-631-51G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-631-5250MG Supplier Sigma Aldrich Supplier No. 366315250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tris(Dibenzylideneacetone)Dipalladium(0)-Chloroform Adduct

Specifications

CAS 52522-40-4
Melting Point 131°C to 135°C
Molecular Formula C52H43Cl3O3Pd2
Linear Formula (C6H5CH=CHCOCH=CHC6H5)3Pd2 -+ CHCl3
MDL Number MFCD00075479
Quantity 250 mg
Synonym Dipalladium-tris(dibenzylideneacetone)chloroform complex; Pd2(dba)3 -+ CHCl3
Molecular Weight (g/mol) 1035.1 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.