Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tris(diethylamido)(tert-butylimido)tantalum(V)

Catalog No. 66899010G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-899-010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 66-899-010G Supplier Sigma Aldrich Supplier No. 66899010G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Atomic number of base material: 73 TantalumPrecursors Packaged for Depositions Systems

Specifications

CAS 169896-41-7
Density 1.252 g/mL (at 25°C)
Molecular Formula C16H39N4Ta
Linear Formula (CH3)3 CNTa(N(C2H5)2)3
Quantity 10 g
MDL Number MFCD02684506
Synonym TBTDET; Tris(diethylamino)(tert-butylimino)tantalum(V)
Molecular Weight (g/mol) 468.46
Percent Purity ≥99.99% (trace metals analysis)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.