Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tris(Dimethylamino)Phosphine

Catalog No. 3932905ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-329-05ML 5 mL
39-329-025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 39-329-05ML Supplier Sigma Aldrich Supplier No. 3932905ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tris(dimethylamino)phosphine can be used as a:Ligand in the preparation of arene-ruthenium(II) complex which is used as a catalyst in hydration of nitriles to amides.Phosphorus source along with InCl3 in the synthesis of InP colloidal quantum dots (QDs).Deoxygenating agent for some +a-dicarbonyl compounds in the presence of fullerene C60.

Specifications

CAS 1608-26-0
Density 0.898 g/mL at 25°C
Boiling Point 48°C to 50°C (12 mmHg, lit.)
Molecular Formula C6H18N3P
Linear Formula P[N(CH3)2]3
Refractive Index n20/D 1.463
MDL Number MFCD00008301
Quantity 5 mL
Synonym (Me2N)3P; Hexamethyltriamidophosphite; Hexamethyltriaminophosphine; HMPT; Hexamethylphosphorous triamide
Molecular Weight (g/mol) 163.2 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.