Learn More
Crescent Chemical Co Inc TRIS(DIMETHYLAMINOMETHYL)PHENO
SDP
Catalog No. 50248216
Encompass_Preferred
Tris(dimethylaminomethyl)phenol pract. Accelerator for epoxy polymerization. Size - 100ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 36975.01 CAS 90-72-2
Catalog No. 50-248-216 Supplier Crescent Chemical Co Inc Supplier No. SE36975.01
Tris(dimethylaminomethyl)phenol pract. Accelerator for epoxy polymerization. Size - 100ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 36975.01 CAS 90-72-2