Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TE Buffer, Tris-EDTA, 1X Solution, pH 7.6, Molecular Biology, Fisher BioReagents

Catalog No. p-29160 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP2474100 100 mL Poly Bottle
BP24741 1 L Nalgene Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP2474100
  • Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X TE buffer solution
  • Eliminates the hassle of dilution or waiting for powder to dissolve
  • 1X solution contains 10mM tris and 1mM EDTA
  • Filtered through a 5 micron filter and autoclaved
  • Tested for the absence of DNase, RNase, and protease
  • TE buffer is routinely used for resuspending nucleic acid samples
  • Intended for laboratory research use only
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris-EDTA
Name Note 1X Solution, pH 7.6
CAS 77-86-1, 6381-92-6, 7647-01-0, 7732-18-5
Color Colorless
pH 7.4 to 8.1
Physical Form Liquid
Quantity 100 mL
DNase DNase free
Filtered Through Filtered through a 5-micron filter.
Packaging Poly Bottle
Protease Protease free
Synonym TE
Grade Molecular Biology
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.