Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TE Buffer, Tris-EDTA, 10X Solution, pH 7.6, Molecular Biology, Fisher BioReagents™

Catalog No. bp24751 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Nalgene Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP24751 1 L Nalgene Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP24751

TE Buffer, Tris-EDTA, 10X Solution, pH 7.6, Molecular Biology

  • Save time and simplify your buffer preparation step by using Fisher BioReagents 10X TE buffer solution - simply dilute as needed
  • 10X solution contains 100mM tris and 10mM EDTA
  • To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TE solution with 9 volumes of ultrapure water, such as BP2819
  • Filtered through a 5 micron filter and autoclaved
  • Tested for the absence of DNase, RNase, and protease
  • TE buffer is routinely used for resuspending nucleic acid samples
  • Intended for laboratory research use only
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris-EDTA (TE), 10X Solution
Name Note pH 7.6
CAS 77-86-1, 6381-92-6, 7647-01-0, 7732-18-5
Color Colorless
pH 7.4 to 8.1
Physical Form Liquid
Quantity 1 L
DNase DNase free
Filtered Through Filtered through a 5-micron filter and autoclaved.
Packaging Nalgene Poly Bottle
Protease Protease free
Synonym TE
Grade Molecular Biology
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.