Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Tris (Hydroxymethyl) Aminomethane Free Base, Reagent 100 GR
SDP

Catalog No. 50980235
Encompass_Preferred

CAS #77-86-1 C₄H₁₁NO₃ F.W. 121.14
Assay: >99.9%

White crystalline powder. A 40% (w/v) aqueous solution. Hydroxymethyl Aminomethane.
Grade: Reagent

Catalog No. 50-980-235 Supplier Electron Microscopy Sciences Supplier No. 11700
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS #77-86-1 C₄H₁₁NO₃ F.W. 121.14
Assay: >99.9%

White crystalline powder. A 40% (w/v) aqueous solution. Hydroxymethyl Aminomethane.
Grade: Reagent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.