Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIS GenAR, Macron Fine Chemicals™

Catalog No. MK7732500 Shop All Avantor Products
Change view
Click to view available options
Quantity:
5 kg
6 x 500 g
Packaging:
POLYSTORMOR
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-7732-500 6 x 500 g POLYSTORMOR
MK7732-5 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MK-7732-500 Supplier Avantor Supplier No. 773204

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Quantity 6 x 500 g
Packaging POLYSTORMOR
Formula Weight 121.14
Chemical Name or Material TRIS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.