Learn More
Kd Medical Inc TRIS GLYC SDS BUFFER, 10X, 4L
SDP
Catalog No. 501031442
Encompass
10X Tris Glycine SDS is used for electrophoretic transfer of protein from polyacrylamide gels to nitrocellulose sheets. One 4 liter cubitainer.
Catalog No. 50-103-1442 Supplier Kd Medical Inc Supplier No. RGH0390
10X Tris Glycine SDS is used for electrophoretic transfer of protein from polyacrylamide gels to nitrocellulose sheets. One 4 liter cubitainer.