Learn More
Research Products International Corp Tris Glycine Buffer 10x Powdr
SDP
Catalog No. 502979110
Encompass_Preferred
Dry homogeneous blend of molecular biology grade TRIS base and Glycine. Add 174.4g to make 1L of 10x Concentrate.
Catalog No. 50-297-9110 Supplier Research Products International Corp Supplier No. T2904010.0
Dry homogeneous blend of molecular biology grade TRIS base and Glycine. Add 174.4g to make 1L of 10x Concentrate.