Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Tris-glycine Native Running Buffer

Catalog No. 04822521 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-822-521 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-822-521 Supplier MP Biomedicals™ Supplier No. 04822521
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tris-glycine Native Running Buffer is a running buffer for protein electrophoresis.

  • Premixed liquid 10X concentrate.
  • Contains 0.25 M TRIS base, 1.92 M Glycine, pH 8.5.
  • Provides excellent separation of native large molecular weight proteins on TRIS-glycine polyacrylamide gels.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris-glycine
Quantity 1 L
Grade Electrophoresis

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.