Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals TRIS-glycine native sample buffer (4X), pH 8.6

Catalog No. AAJ61864AD
Change view
Click to view available options
Quantity:
25 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61864AD 50 mL
AAJ61864AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61864AD Supplier Thermo Scientific Chemicals Supplier No. J61864AD
Add to Cart

missing translation for 'validMainframeMsg'

For preparation of protein samples for native gel electrophoresis

For preparation of protein samples for native gel electrophoresis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sample Loading Buffer
Color Blue
Concentration 4X
For Use With (Application) Preparation of protein samples for native gel electrophoresis
pH 8.6
Physical Form Liquid
Quantity 50 mL

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.