Learn More
Bioworld Tris-Glycine Sample Buffer (4X), 50 mL
Catalog No. 50198651
Encompass_Preferred
Product DescriptionTris-Glycine (TG) sample buffer has a concentration of 4X. It contains 400 mM Tris-HCl at a pH of 8.6, 40% glycerol, and 0.02% bromophenol blue.TG buffers are blended using pure reagents to provide high quality for protein research. This buffer is formulated to ensure exceptional purity along with the absence of nuclease and protease activities. TG buffer is widely used for electroblotting proteins in Western blot procedures and is recommended for use with all nitrocellulose and nylon membranes.
Catalog No. 50-198-651 Supplier Bioworld Supplier No. 214200222
Add to Cart
Add to Cart
Product DescriptionTris-Glycine (TG) sample buffer has a concentration of 4X. It contains 400 mM Tris-HCl at a pH of 8.6, 40% glycerol, and 0.02% bromophenol blue.TG buffers are blended using pure reagents to provide high quality for protein research. This buffer is formulated to ensure exceptional purity along with the absence of nuclease and protease activities. TG buffer is widely used for electroblotting proteins in Western blot procedures and is recommended for use with all nitrocellulose and nylon membranes.