Learn More
Research Products International Corp Tris-Glycine-SDS, 10 L
SDP
Catalog No. 501977350
Encompass_Preferred
Common buffer used for running SDS-PAGE gels in electrophoresis.
1X Solution Contains:
TRIS/TRIS HCl     25 mM
Glycine               0.192 M
SDS                 0.1%(w/v)
Catalog No. 50-197-7350 Supplier Research Products International Corp Supplier No. T3208010000.0
Common buffer used for running SDS-PAGE gels in electrophoresis.
1X Solution Contains:
TRIS/TRIS HCl     25 mM
Glycine               0.192 M
SDS                 0.1%(w/v)