Learn More
Research Products International Corp TRIS-Glycine-SDS Buffer, 10X Powder, 184 Grams of Powder, Makes 1 Liter of Solution
SDP
Catalog No. 501368632
Encompass_Preferred
Composition:
Glycine: < 80.0%
TRIS (Hydroxymethyl) Aminomethane: < 20.0%
SDS: < 6.0%
Catalog No. 50-136-8632 Supplier Research Products International Corp Supplier No. T280201.0
Composition:
Glycine: < 80.0%
TRIS (Hydroxymethyl) Aminomethane: < 20.0%
SDS: < 6.0%