Learn More
Bioworld Tris-Glycine-SDS (TGS) Running Buffer (10X), 1 L
Catalog No. 50198652
Encompass_Preferred
Product DescriptionTris-Glycine SDS (TGS) running buffer has a concentration of 10X with a pH of 8.3. 1X solution contains 0.025M Tris, 0.192M Glycine, and 0.1% SDS. Product is of ultra pure grade.TG buffers are blended using pure reagents to provide high quality for protein research. This buffer is formulated to ensure exceptional purity along with the absence of nuclease and protease activities. TG buffer is widely used for electroblotting proteins in Western blot procedures and is recommended for use with all nitrocellulose and nylon membranes.
Catalog No. 50-198-652 Supplier Bioworld Supplier No. 214200231
Product DescriptionTris-Glycine SDS (TGS) running buffer has a concentration of 10X with a pH of 8.3. 1X solution contains 0.025M Tris, 0.192M Glycine, and 0.1% SDS. Product is of ultra pure grade.TG buffers are blended using pure reagents to provide high quality for protein research. This buffer is formulated to ensure exceptional purity along with the absence of nuclease and protease activities. TG buffer is widely used for electroblotting proteins in Western blot procedures and is recommended for use with all nitrocellulose and nylon membranes.