Learn More
Sigma Aldrich Fine Chemicals Biosciences Tris-Glycine-SDS Buffer 101L
Catalog No. 502900053
Encompass
Tris-glycine-SDS (TGS) running buffer is the most commonly used buffer for sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE) of proteins. TGS is usually used for both anode and cathode buffer. Recommended running conditions is 150 volts for mini vertical gel electrophoresis units.
Catalog No. 50-290-0053 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T77771L
Tris-glycine-SDS (TGS) running buffer is the most commonly used buffer for sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE) of proteins. TGS is usually used for both anode and cathode buffer. Recommended running conditions is 150 volts for mini vertical gel electrophoresis units.