Learn More
Kd Medical Inc TRIS-GLYCINE-SDS BUFFER 10X
SDP
Catalog No. 501018898
Encompass_Preferred
Tris-Glycine-SDS PAGE Buffer (10X) is a concentrated solution of Tris-Glycine-SDS in distilled/deionized water, especially intended for use as the running buffer in SDS-PAGE of proteins. One 1 liter bottle.
Catalog No. 50-101-8898 Supplier Kd Medical Inc Supplier No. RGF3390
Tris-Glycine-SDS PAGE Buffer (10X) is a concentrated solution of Tris-Glycine-SDS in distilled/deionized water, especially intended for use as the running buffer in SDS-PAGE of proteins. One 1 liter bottle.