Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris-Glycine-SDS Buffer, 10X Solution, Molecular Biology Grade, Hoefer™
SDP

Catalog No. 03500532
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-500-532 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-500-532 Supplier Hoefer, Inc Supplier No. GR1491
May include imposed supplier surcharges.
Add to Cart
Add to Cart

For PAGE, dilute to 1X with high purity H2O for a solution consisting of 25mM Tris, pH 8.3, 192mM glycine and 0.1% SDS

Use in PAGE and blotting

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Quantity 1 L
Filtered Through 0.2μm filtered
Concentration or Composition (by Analyte or Components) 0.25M Tris, 1.92 M glycine and 1.0% SDS in high purity H20
Chemical Name or Material Tris-Glycine-SDS Buffer, 10X Solution, pH 8.3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.