Learn More
National Diagnostics TRIS-GLYCINE-SDS PAGE BUFFER (
SDP
Catalog No. 5089990103
Encompass
Tris-Glycine-SDS PAGE Buffer (10X), Ultra-pure concentrated standard running buffer for SDS-PAGE. Formulated with 18 MegOhm water. 0.2 micron filtration, 4 LT
Catalog No. 50-899-90103 Supplier National Diagnostics Supplier No. EC8704LTR
Add to Cart
Add to Cart
Tris-Glycine-SDS PAGE Buffer (10X), Ultra-pure concentrated standard running buffer for SDS-PAGE. Formulated with 18 MegOhm water. 0.2 micron filtration, 4 LT