Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boston Bioproducts Inc Tris-Glycine-SDS Running Buffer 10X, 4 L, 8.50 ± 0.20 pH, 250 mM Tris, 1.92 M Glycine, 1% SDS, Anode and Cathode buffer, SDS-PAGE, Western blotting

Catalog No. NC9163508
Encompass

This Tris-Glycine SDS Running Buffer (10X) is specifically formulated for use in protein electrophoresis applications, such as SDS-PAGE and Western blotting. It plays a vital role in the separation and resolution of denatured proteins. Upon dilution to the recommended working concentration, the buffer ensures consistent results with well-resolved protein bands. Furthermore, this buffer can function effectively as both the anode and cathode buffer, maintaining the pH essential for optimum electrophoresis performance.

  • Volume: 4 L
  • Concentration: 10X
  • pH: 8.50 ± 0.20
  • Application: SDS-PAGE, Western blotting
  • Function: Separation of denatured proteins
  • Recommended dilution: 10-fold with deionized water for 1X use
  • Storage conditions: Room temperature
  • Filtered through: 1-micron filter
  • Quality control testing: Appearance, pH

Catalog No. NC9163508 Supplier Boston Bioproducts Inc Supplier No. BP1504L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Tris-Glycine SDS Running Buffer (10X) is specifically formulated for use in protein electrophoresis applications, such as SDS-PAGE and Western blotting. It plays a vital role in the separation and resolution of denatured proteins. Upon dilution to the recommended working concentration, the buffer ensures consistent results with well-resolved protein bands. Furthermore, this buffer can function effectively as both the anode and cathode buffer, maintaining the pH essential for optimum electrophoresis performance.

  • Volume: 4 L
  • Concentration: 10X
  • pH: 8.50 ± 0.20
  • Application: SDS-PAGE, Western blotting
  • Function: Separation of denatured proteins
  • Recommended dilution: 10-fold with deionized water for 1X use
  • Storage conditions: Room temperature
  • Filtered through: 1-micron filter
  • Quality control testing: Appearance, pH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.