Learn More
Bioworld Tris-Glycine Transfer Buffer (10X), 1 L
Catalog No. 50198550
Encompass_Preferred
Tris-Glycine Transfer Buffer, 10X Concentrate.For Western blotting and gel electrophoresis. Tris-glycine buffer is used to make a Tris-glycine-methanol transfer buffer, which is the most common protein transfer buffer for wet blot transfers. The methanol prevents the gel from swelling during the transfer and enhances the protein binding to nitrocellulose.The 10× Tris-glycine buffer is diluted to 1× with methanol and water to make a solution containing 25 mM Tris, 192 mM glycine, and 20% methanol. A sufficient amount of transfer buffer should be made to cover the electrode wires in the wet blot transfer unit and to soak the gel, membrane and blotting paper.To make 1X Transfer Buffer from 10X concentrate: Mix 100 ml of 10X Transfer Buffer, 200 ml of methanol and 700 ml of dd water per liter.Foreign activity: Protease, none detected Storage temp.: 2-8°C
Catalog No. 50-198-550 Supplier Bioworld Supplier No. 209600061
Tris-Glycine Transfer Buffer, 10X Concentrate.For Western blotting and gel electrophoresis. Tris-glycine buffer is used to make a Tris-glycine-methanol transfer buffer, which is the most common protein transfer buffer for wet blot transfers. The methanol prevents the gel from swelling during the transfer and enhances the protein binding to nitrocellulose.The 10× Tris-glycine buffer is diluted to 1× with methanol and water to make a solution containing 25 mM Tris, 192 mM glycine, and 20% methanol. A sufficient amount of transfer buffer should be made to cover the electrode wires in the wet blot transfer unit and to soak the gel, membrane and blotting paper.To make 1X Transfer Buffer from 10X concentrate: Mix 100 ml of 10X Transfer Buffer, 200 ml of methanol and 700 ml of dd water per liter.Foreign activity: Protease, none detected Storage temp.: 2-8°C