Learn More
Boston Bioproducts Inc Tris-HCl Buffer (1 M, pH 8.0, RNase Free) - 250ml
Catalog No. NC2045852
Encompass
The Tris-HCl Buffer is a widely used bufering reagent in molecular biology and biochemistry, acting to maintain a stable pH, and preserve the integrity of target macromolecules. The buffering capacity of Tris-HCl is effective in the pH range of 7.0 to 9.0, making it ideal for many biological applications like cell culture, molecular biology and DNA/RNA extraction,. This buffer is also tested for RNase enzymes, minimizing the risk of nucleic acid degredation in later applications.
Catalog No. NC2045852 Supplier Boston Bioproducts Inc Supplier No. BBT80DR250ML
Add to Cart
Add to Cart
The Tris-HCl Buffer is a widely used bufering reagent in molecular biology and biochemistry, acting to maintain a stable pH, and preserve the integrity of target macromolecules. The buffering capacity of Tris-HCl is effective in the pH range of 7.0 to 9.0, making it ideal for many biological applications like cell culture, molecular biology and DNA/RNA extraction,. This buffer is also tested for RNase enzymes, minimizing the risk of nucleic acid degredation in later applications.