Learn More
Gold Biotechnology Inc Tris HCl 1 kg
SDP
Catalog No. NC2943382
Encompass_Preferred
Short for Tris (hydroxymethyl) aminomethane (THAM) hydrochloride, Tris HCl is an organic compound often used in buffer solutions such as TAE or TBE for electrophoresis gels. Tris is highly soluble in water and is useful in the pH range 7.0-9.0. It is used in the preparation of Laemmli buffer, one of the most common SDS-PAGE buffers. In addition, Tris can also be used for many custom running and loading buffers, most often with glycine and SDS. The use of single-junction pH electrodes that contain silver should be avoided when determining the pH of a solution containing Tris buffer.
Catalog No. NC2943382 Supplier Gold Biotechnology Inc Supplier No. T0951
Short for Tris (hydroxymethyl) aminomethane (THAM) hydrochloride, Tris HCl is an organic compound often used in buffer solutions such as TAE or TBE for electrophoresis gels. Tris is highly soluble in water and is useful in the pH range 7.0-9.0. It is used in the preparation of Laemmli buffer, one of the most common SDS-PAGE buffers. In addition, Tris can also be used for many custom running and loading buffers, most often with glycine and SDS. The use of single-junction pH electrodes that contain silver should be avoided when determining the pH of a solution containing Tris buffer.