Learn More
Kd Medical Inc TRIS HCL, 1M PH 6.8, 500ML
SDP
Catalog No. 501031384
Encompass_Preferred
Tris, Hydrochloride is commonly used as a buffer component for phenol extractions of DNA or RNA and a buffering component of separating and stacking gels in the characterization of protein products by SDS-PAGE. 1M solution, pH 6.8. One 500ml bottle.
Catalog No. 50-103-1384 Supplier Kd Medical Inc Supplier No. RGE3355
Tris, Hydrochloride is commonly used as a buffer component for phenol extractions of DNA or RNA and a buffering component of separating and stacking gels in the characterization of protein products by SDS-PAGE. 1M solution, pH 6.8. One 500ml bottle.