Learn More
Kd Medical Inc TRIS-HCL, 1M, PH 7.4, 500ML
SDP
Catalog No. 501031382
Encompass_Preferred
Tris, Hydrochloride is commonly used as a buffer component for phenol extractions of DNA or RNA and a buffering component of separating and stacking gels in the characterization of protein products by SDS-PAGE. 1M solution, pH 7.4. One 500ml bottle.
Catalog No. 50-103-1382 Supplier Kd Medical Inc Supplier No. RGE3340
Add to Cart
Add to Cart
Tris, Hydrochloride is commonly used as a buffer component for phenol extractions of DNA or RNA and a buffering component of separating and stacking gels in the characterization of protein products by SDS-PAGE. 1M solution, pH 7.4. One 500ml bottle.