Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Tris-HCl (1M), pH 7.5, 500 mL
SDP

Catalog No. 507517339
Encompass_Preferred

Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. These solutions of Tris Hydrochloride provide convenient and highly purified solutions. Applications: Tris solutions are formulated into working concentrations for preparing the stacking and resolving portions of native or SDS-PAGE gels using discontinuous buffer systems according to Laemmli (1970) or Ornstein and Davis (1959).

Use Use Tris-HCl (1M), pH 7.0-7.5 for DEAE Dextran-mediated transfection of non-adherent and adherent cells.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
DNase, RNase and Protease Tested
pH Range: 7.5 ± 0.1
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-751-7339 Supplier Quality Biological Inc Supplier No. 351006101
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. These solutions of Tris Hydrochloride provide convenient and highly purified solutions. Applications: Tris solutions are formulated into working concentrations for preparing the stacking and resolving portions of native or SDS-PAGE gels using discontinuous buffer systems according to Laemmli (1970) or Ornstein and Davis (1959).

Use Use Tris-HCl (1M), pH 7.0-7.5 for DEAE Dextran-mediated transfection of non-adherent and adherent cells.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
DNase, RNase and Protease Tested
pH Range: 7.5 ± 0.1
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.