Learn More
Kd Medical Inc TRIS HCL, 1M, PH 7.5, 500ML
SDP
Catalog No. 501031383
Encompass_Preferred
Tris, Hydrochloride is commonly used as a buffer component for phenol extractions of DNA or RNA and a buffering component of separating and stacking gels in the characterization of protein products by SDS-PAGE. 1M solution, pH 7.5. One 500ml bottle.
Catalog No. 50-103-1383 Supplier Kd Medical Inc Supplier No. RGE3350
Tris, Hydrochloride is commonly used as a buffer component for phenol extractions of DNA or RNA and a buffering component of separating and stacking gels in the characterization of protein products by SDS-PAGE. 1M solution, pH 7.5. One 500ml bottle.