Learn More
Quality Biological Inc Tris HCL (1 M), pH 8.0, 500 mL
SDP
Catalog No. 507517340
Encompass_Preferred
A hybridization buffer specifically formulated for use with nylon membranes. ¿ It does not contain Denhardt¿s Reagent, which may interfere with the hybridization reaction of DNA immobilized on nylon. ¿ Formulated for Nylon membrane ¿ Filtered through a 0.22 ¿m membrane ¿ DNase, RNase, and Protease tested ¿ Applications: Denatured fragmented salmon sperm DNA may be added to this hybridization buffer to reduce nonspecific background. ¿ Storage Conditions: 25¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ DNase Tested ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 36 months
Catalog No. 50-751-7340 Supplier Quality Biological Inc Supplier No. 351007101
Add to Cart
Add to Cart
A hybridization buffer specifically formulated for use with nylon membranes. ¿ It does not contain Denhardt¿s Reagent, which may interfere with the hybridization reaction of DNA immobilized on nylon. ¿ Formulated for Nylon membrane ¿ Filtered through a 0.22 ¿m membrane ¿ DNase, RNase, and Protease tested ¿ Applications: Denatured fragmented salmon sperm DNA may be added to this hybridization buffer to reduce nonspecific background. ¿ Storage Conditions: 25¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ DNase Tested ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 36 months