Learn More
Kd Medical Inc TRIS HCL 1M PH 8.5, 500ML
SDP
Supplier:  Kd Medical Inc RGE3363
Encompass_Preferred
Tris, Hydrochloride is commonly used as a buffer component for phenol extractions of DNA or RNA and a buffering component of separating and stacking gels in the characterization of protein products by SDS-PAGE. 1M solution, pH 8.5. One 500ml bottle.
Catalog No. 50-103-1386