Learn More
Kd Medical Inc TRIS HCL 1M PH 8.O 500ML
SDP
Catalog No. 501018922
Encompass_Preferred
Tris, Hydrochloride is commonly used as a buffer component for phenol extractions of DNA or RNA and a buffering component of separating and stacking gels in the characterization of protein products by SDS-PAGE. 1M solution, pH 8.0. One 500ml bottle.
Catalog No. 50-101-8922 Supplier Kd Medical Inc Supplier No. RGE3360
Add to Cart
Add to Cart
Tris, Hydrochloride is commonly used as a buffer component for phenol extractions of DNA or RNA and a buffering component of separating and stacking gels in the characterization of protein products by SDS-PAGE. 1M solution, pH 8.0. One 500ml bottle.