Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Tris (1M), pH 9.8 1000 mL
SDP

Catalog No. 502209779
Encompass_Preferred

Suitable for molecular biology protein chemistry and biochemical applications.

DNase RNase and Protease tested.
Filtered through a 0.22 µm membrane

Applications: Tris Buffer (1M), pH 9.8 is commonly used in SDS-PAGE and Electrophoresis applications.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
DNase RNase and Protease Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-220-9779 Supplier Quality Biological Inc Supplier No. 351176131
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for molecular biology protein chemistry and biochemical applications.

DNase RNase and Protease tested.
Filtered through a 0.22 µm membrane

Applications: Tris Buffer (1M), pH 9.8 is commonly used in SDS-PAGE and Electrophoresis applications.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
DNase RNase and Protease Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.