Learn More
Quality Biological Inc TRIS HCL (1 M), pH 7.0, 500 mL
SDP
Catalog No. 507517634
Encompass_Preferred
Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. ¿ Provide convenient and highly purified solutions. ¿ Applications: Tris solutions are formulated into working concentrations for preparing the stacking and resolving portions of native or SDS-PAGE gels.¿ Use Tris-HCl 1 M pH 7.0-7.5 for DEAE Dextran-mediated transfection of non-adherent and adherent cells. ¿ Storage Conditions: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ DNase RNase and Protease Tested ¿ pH: 7.5¿ 0.1 ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 36 months
Catalog No. 50-751-7634 Supplier Quality Biological Inc Supplier No. 351050101
Add to Cart
Add to Cart
Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. ¿ Provide convenient and highly purified solutions. ¿ Applications: Tris solutions are formulated into working concentrations for preparing the stacking and resolving portions of native or SDS-PAGE gels.¿ Use Tris-HCl 1 M pH 7.0-7.5 for DEAE Dextran-mediated transfection of non-adherent and adherent cells. ¿ Storage Conditions: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ DNase RNase and Protease Tested ¿ pH: 7.5¿ 0.1 ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 36 months