Learn More
Gold Biotechnology Inc Tris HCl 500 g
SDP
Catalog No. 5000562127
Encompass_Preferred
Tris hydrochloride is a reagent that is used for the stabilization of buffer formulations and electrophoresis of biological molecules
Catalog No. 50-005-62127 Supplier Gold Biotechnology Inc Supplier No. T095500
Add to Cart
Add to Cart
Tris hydrochloride is a reagent that is used for the stabilization of buffer formulations and electrophoresis of biological molecules