Learn More
Sigma Aldrich Fine Chemicals Biosciences Trizma hydrochloride, 1185-53-1, MFCD00012590, 500 g
Catalog No. NC0619460
Encompass
Linear Formula: NH2C(CH2OH)3 · HCl, Molecular Weight: 157.60, BioPerformance Certified, cell culture tested, ≥99.0% (titration), Synonym: TRIS HCl, TRIS hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride.
Catalog No. NC0619460 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T5941500G
Linear Formula: NH2C(CH2OH)3 · HCl, Molecular Weight: 157.60, BioPerformance Certified, cell culture tested, ≥99.0% (titration), Synonym: TRIS HCl, TRIS hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride.