Learn More
Biotang Inc TRIS-HCL Buffer 1.0M, pH 7.5 (0.2 micron filtered), 500 ml
Catalog No. 50155715 Shop All BIOTANG Inc Products
Encompass_Preferred
TRIS-HCL Buffer 1.0M, pH 7.5 (0.2 micron filtered), 500 ml.
Catalog No. 50-155-715 Supplier Biotang Inc Supplier No. BTBB165
TRIS-HCL Buffer 1.0M, pH 7.5 (0.2 micron filtered), 500 ml.