Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotang Inc TRIS-HCL Buffer 1.0M, pH 8.5, 1000ml

Supplier:  Biotang Inc BTBB565

Encompass

TRIS-HCL Buffer 1.0M, pH 8.5, 1000 ml.

Catalog No. 50-155-887


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.