Learn More
Bioworld Tris-HCl Buffer 10mM, pH 8.5, 500ml
Catalog No. NC1213695
Encompass_Preferred
Common buffer for biological sciences. Technical Information: Biotechnology grade, pH 8.5 +-0.1. Warnings: Causes irritation to eyes, skin, and mucus membranes. Do not inhale or ingest. Wash thoroughly after handling. Appearance: Liquid, PH Value: 8.5, Molecular Weight : 157.6, Molecular Formula : C4H11NO3.HCl, pH : 8.5, Solubility : Soluble in water. Appearance Form : Liquid. Appearance Color : Clear. 500ml
Catalog No. NC1213695 Supplier Bioworld Supplier No. 420204141
Common buffer for biological sciences. Technical Information: Biotechnology grade, pH 8.5 +-0.1. Warnings: Causes irritation to eyes, skin, and mucus membranes. Do not inhale or ingest. Wash thoroughly after handling. Appearance: Liquid, PH Value: 8.5, Molecular Weight : 157.6, Molecular Formula : C4H11NO3.HCl, pH : 8.5, Solubility : Soluble in water. Appearance Form : Liquid. Appearance Color : Clear. 500ml