Learn More
Bioworld Tris-HCl Buffer 1M, pH 7.8 | 1L
Supplier:  Bioworld 420202761
Encompass_Preferred
Tris-HCl Buffer 1M, pH 7.8 is a stabilizing buffer with various biological applications in electrochromatography, UV analysis, and HPLC. It is used to adjust and stabilize pH ranges for gels used in gel electrophoresis applications.
Catalog No. NC1456433