Learn More
Abcam Tris-HCl (pH 8.8), 100ML
Catalog No. 502205753
Encompass_Preferred
A 1.5 M solution of Tris HCl in water (pH 8.8).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5753 Supplier Abcam Supplier No. AB286854100ML
A 1.5 M solution of Tris HCl in water (pH 8.8).
The product is subject to the following: Abcam Restricted Use Statement