Learn More
AG Scientific Inc TRIS Hydrochloride, 500 G
Catalog No. 502237052
Encompass_Preferred
Tris-HCl is a high purity biological buffer. Pre-weighed based on molar weight instead of grams in ready-to-use pouches. Stock solutions easy and fast to prepare. DNase, RNase and Proteases not detected. Ready-to-use. Prepare stock solution without weighing the dry product. Moisture proof pouches ensure high quality and ease of use. Purity data is for a dried basis.CAS Number: 1185-53-1Molecular Weight: 157.6 DaChemical Formula: C4H11NO3HClSolubility: Water: clear colorless solutionStorage Temperature: Room TemperatureResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-7052 Supplier AG Scientific Inc Supplier No. T1176500GM
Tris-HCl is a high purity biological buffer. Pre-weighed based on molar weight instead of grams in ready-to-use pouches. Stock solutions easy and fast to prepare. DNase, RNase and Proteases not detected. Ready-to-use. Prepare stock solution without weighing the dry product. Moisture proof pouches ensure high quality and ease of use. Purity data is for a dried basis.CAS Number: 1185-53-1Molecular Weight: 157.6 DaChemical Formula: C4H11NO3HClSolubility: Water: clear colorless solutionStorage Temperature: Room TemperatureResearch or further manufacturing use only, not for food or drug use.