Learn More
Pfanstiehl Inc Tris Hydrochloride (Tris HCl) High Purity, Low Endotoxin, Low Metals (T-154) 5kg GMP CAS: 1185-53-1 Injectable Excipient Grade
Catalog No. NC2953483 Shop All Pfanstiehl Products
Encompass
Pfanstiehl Tris HCL, High Purity, Low Endotoxin, Low Metals, is a highly purified buffer component that has been specifically designed and formulated to remove impurities such as endotoxin, trace metals, and nitrosamines. Manufactured in the USA under ICH-Q7 cGMP conditions, Pfanstiehl Tris HCl meets parenteral (injectable) excipient grade standards, making it ideal for pharmaceutical, biologic, and vaccine bioprocessing and formulation. cGMP , pharmaceutical grade .
Catalog No. NC2953483 Supplier Pfanstiehl Inc Supplier No. T1545KG
Pfanstiehl Tris HCL, High Purity, Low Endotoxin, Low Metals, is a highly purified buffer component that has been specifically designed and formulated to remove impurities such as endotoxin, trace metals, and nitrosamines. Manufactured in the USA under ICH-Q7 cGMP conditions, Pfanstiehl Tris HCl meets parenteral (injectable) excipient grade standards, making it ideal for pharmaceutical, biologic, and vaccine bioprocessing and formulation. cGMP , pharmaceutical grade .