Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIS hydrochloride, >99%, Cell culture reagent grade, MP Biomedicals™

Catalog No. 0219455891 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-194-55891 1 kg
ICN19455880 100 g
ICN19455890 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-194-55891 Supplier MP Biomedicals, Inc Supplier No. 0219455891
Add to Cart
Add to Cart

Excellent biochemical and biological buffer for all applications where ultra high purity is required. The pH values of all buffers are temperature and concentration dependent

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
MDL Number MFCD00012590
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
Synonym Tris(hydroxymethyl)aminomethane hydrochloride, TRIS HCl
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Color White
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range > 99%
Formula Weight 157.6
Grade Cell Culture
Chemical Name or Material TRIS hydrochloride
EINECSNumber 214-684-5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.